Vanadium standard solution, 1000 mg/L V in dil. nitric acid standard for AAS
Catalog # PROOU-IAA-223
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:Nitric acid dil.
- Pack type:Glass bottle
- Pk:125 mL
- Element:Vanadium
- Symbol:V