Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Terbium, Sigma-Aldrich®

Terbium, Sigma-Aldrich®

Glass bottle for solids, 5 g
Catalog # SIAL263206-5G
Supplier:   Merck
Terbium, Sigma-Aldrich®
Catalog # SIAL263206-5G
Supplier:  Merck
CAS Number:  7440-27-9
Terbium, Sigma-Aldrich®

Glass bottle for solids, 5 g

CAS Number:  7440-27-9
Price
Inquire For Price
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:7440-27-9
  • Formula:Tb
  • MDL Number:MFCD00011256
  • Molecular Weight:158.93 g/mol
  • Merck Index:12,09300
  • Boiling Pt:3041 °C (1013 hPa)
  • Melting Pt:1356 °C
  • Density:8.27 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ100
  •  :Density: 8.27 g/cm³ (20 °C)
  • EINECS:231-137-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.