Order Entry
Slovenia
ContactUsLinkComponent
Lutetium standard solution, 1000 mg/L Lu in dil. nitric acid standard for ICP
Lutetium standard solution, 1000 mg/L Lu in dil. nitric acid standard for ICP
  PROOU-ICP-071
 :  
Lutetium standard solution, 1000 mg/L Lu in dil. nitric acid standard for ICP
  PROOU-ICP-071
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    125 mL
  • Element:
    Lutetium
  • Symbol:
    Lu