Arsenic standard solution, 10,000 mg/L As in dil. nitric acid standard for ICP
Catalog # PROOU-ICP-133
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Concentration:10000 mg/L
- Pack type:Glass bottle
- Pk:125 mL
- Element:Arsenic
- Symbol:As