Tin standard solution, 1000 mg/L Sn in dil. nitric acid standard for ICP
Catalog # PROOU-ICP-050
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:Nitric acid dil.
- Pack type:Glass bottle
- Pk:125 mL
- Element:Tin
- Symbol:Sn