Lead
Glass bottle for solids, 500 mg
Catalog # PROOLGC2608
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
- CAS Number:7439-92-1
- Formula:Pb
- MDL Number:MFCD00134050
- Molecular Weight:207.2 g/mol
- Merck Index:12,05410
- Boiling Pt:1740 °C (1013 hPa)
- Melting Pt:327 °C
- Density:11.3437 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-100-4
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg