Microcentrifuge, refrigerated, 5424 R (General Lab Product)
Supplier: EPPENDORF
Certificates
About this item
This microcentrifuge with 24 positions is ideal for everyday laboratory use. Available in two versions: with rotary control knobs or touch sensitive keypad.
- Rotary knob version for fast parameter selection
- Keypad version for easy cleaning
- Soft-touch one-finger lid closure for ergonomic operation
- OptiBowl® design for quiet operation, even without a rotor lid
- FastTemp function for quick pre-cooling of the centrifuge, e.g. from 21 °C to 4 °C in only 8 minutes
- ECO power shut-off engages after 8 hours of non-use to reduce energy consumption (40 % over night) and extend compressor life (can be deactivated)
- Built-in condensation drain to eliminate water accumulation in the rotor chamber
Ordering information: Supplied without rotor or with 24×1,5/2,0 ml standard rotor FA-45-24-11 and fitted lid.