Standards for LC performance, test mix, CRM
Catalog # REST31699
Supplier: Restek
CAS Number:
Specifications
- Pack type:Ampoule
- Pk:1 Ampoul
- Description:LC performance test mix
- Pk Info:1 ml
Specifications
About this item
Highly effective for characterising LC column parameters and tested against the NIST 870 standard.
- Mixture of 5 components in methanol
- Simple, easy, reliable approach to quality control (QC) evaluations or column classification
- Monitor column performance over time
The test mix contains: amitriptyline hydrochloride (549-18-8) 2,800 µg/ml; ethylbenzene (100-41-4) 1,700 µg/ml; quinizarin (81-64-1) 94 µg/ml; toluene (108-88-3) 1,400 µg/ml; uracil (66-22-8) 28 µg/ml.
Certifications: Meets CRM, ISO requirements.