Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Environmental Test Kits
Reagent for nickel determination using Lovibond® MD photometers
Reagent for nickel determination using Lovibond® MD photometers
Reagent for nickel determination using Lovibond® MD photometers
LOVI2419033
:
Lovibond
:
Reagent for nickel determination using Lovibond® MD photometers
LOVI2419033
:
Lovibond
:
Chemical method:
Dimethylglyoxime
Pk:
50 Tests
Description:
Nickel-51, Nickel-52 reagent test
Range:
0.02 - 1 mg/l, 0.2 - 7 mg/l Ni