Mg²⁺/ATP Activating Solution
ENZOBMLEW98050100
:
- Pk:100 µl
- Seq:02
- Enzyme name:Mg2+/ATP activating solution
This solution contains pre-coupled Mg-ATP for immediate use as an energy source for a range of ATP-dependent reactions (e.g. ubiquitin thioester conjugation, proteasomal degradation, SUMO conjugation).
Liquid. In 50mM TRIS-Cl, pH 7,5, containing 0,1 M ATP and 0,1 M MgCl₂.