ContactUsLinkComponent
Z-VVR-AMC
Z-VVR-AMC
Catalog # ENZOBMLP1990010
Supplier:  Enzo Life Sciences
CAS Number:  
Z-VVR-AMC
Catalog # ENZOBMLP1990010
Supplier:  Enzo Life Sciences
CAS Number:  
Specifications
  • Pk:
    10 mg
  • Seq:
    02
  • Protein/peptide name:
    Z-VVR-AMC
  • Purity:
    >99% (TLC)
Specifications
About this item
Substrate for cathepsin S and cathepsin L
Sensitive fluorogenic substrate for cathepsin S and cathepsin L. Also cleaved by cathepsin B, cathepsin V, kallikrein 8, and factor VIIa/TF but not cathepsin K nor kallikrein 2. Ex.: 380 nm, Em.: 460 nm, although the following Ex/Em can also be used: 355,375/440,450. This substrate is useful for inhibitor screening and kinetic analysis. Also available: fluorogenic calibration standard AMC (BML-KI144).