Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Buffers
Assay Buffers
MMP-3 fluorescent assay buffer
MMP-3 fluorescent assay buffer
MMP-3 fluorescent assay buffer
Catalog # ENZOBMLKI1270020
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
MMP-3 fluorescent assay buffer
Catalog # ENZOBMLKI1270020
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
20 mL
Description:
MMP-3 Fluorescent assay buffer
Seq:
02
Specifications
About this item
Product Details
Product Details