Orders LinkContactUsLinkComponent
AMPIGENE® Taq mix
AMPIGENE® Taq mix
Supplier:   Enzo Life Sciences
AMPIGENE® Taq mix
Supplier:  Enzo Life Sciences
Certificates
3 Options Available Below
Product Details & Documents
Optimised standard PCR mix for enhanced speed, yield, and specificity. Used for standard PCR applications.
AMPIGENE® Taq Mix uses the latest developments in polymerase technology and buffer chemistry to optimize PCR. The AMPIGENE® Taq Mix is a robust mix for routine PCR applications including genotyping, screening, and library construction. An optimized buffer system allows efficient amplification under fast and standard cycling conditions.
Delivery information: Kit contains 2×AMPIGENE® Taq Mix (AMPIGENE® Taq Polymerase, 6 mM MgCl₂, 2 mM dNTPs, enhancer and stabilizers).
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • ENZOENZNUC1000040
  • ENZOENZNUC1000200
  • ENZOENZNUC1001000
  • Storage conditions:
  • −20 °C
  • −20 °C
  • −20 °C
Specifications
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • ENZOENZNUC1000040
    AMPIGENE® Taq mix
    AMPIGENE® Taq mix
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 52,80 €
    52,80 €
    1 * 1 KIT
       52,80 €
    Specifications:
    More
    Specifications
    Cat. No.
    ENZOENZNUC1000040
    Storage conditions
    −20 °C
  • ENZOENZNUC1000200
    AMPIGENE® Taq mix
    AMPIGENE® Taq mix
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 170,00 €
    170,00 €
    1 * 1 KIT
       170,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ENZOENZNUC1000200
    Storage conditions
    −20 °C
  • ENZOENZNUC1001000
    AMPIGENE® Taq mix
    AMPIGENE® Taq mix
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 609,00 €
    609,00 €
    1 * 1 KIT
       609,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ENZOENZNUC1001000
    Storage conditions
    −20 °C