Serotonin ELISA Kit
ENZOADI900175
:
- Pk:1 KIT
- Assay Type:Competitive
- Target protein:Serotonin
- Description:Serotonin ELISA kit
- Seq:02
- Storage conditions:–20 °C (long term)
- Sensitivity:0,293 ng/ml (range 0,49 - 500 ng/ml)
The Serotonin EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours.
- Highly sensitive measurement of serotonin
- No acylation required for standards and samples
- Validated in multiple complex sample matrices
- High throughput format with results
- Fully quantitative results that surpass semi-quantitiave Western blot analysis
For the quantitative determination of Serotonin in platelets, plasma, serum, and urine from any species.
: Supplied with GxR IgG microtiter plate, conjugate, antibody, assay buffer concentrate, wash buffer concentrate, standard, pNpp substrate, stop solution