Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Biological Standards
FLUOR DE LYS® deacetylated standard
FLUOR DE LYS® deacetylated standard
FLUOR DE LYS® deacetylated standard
Catalog # ENZOBMLKI1420030
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
FLUOR DE LYS® deacetylated standard
Catalog # ENZOBMLKI1420030
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
30 µl
Description:
FLUOR DE LYS® deacetylated standard, 10 mm, in DMSO
Seq:
02
Specifications
About this item
Product Details
Product Details