Anti-BCMA Rat Monoclonal Antibody [clone: Vicky-1]
ENZOALX804151C100
- Antibody type:Primary
- Antigen name:B-cell maturation antigen
- Antigen symbol:BCMA
- Clonality:Monoclonal
- Clone:Vicky-1
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- Isotype:IgG1
- Western blot:Yes
- Seq:02
- Form:Liquid
- Antigen synonyms:CD269|TNFRSF17|BCMA|BCM
- Amino acid number:2 to 54
- Storage buffer:PBS containing 0,02% sodium azide.
- Storage temperature:–20 °C
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant human BCMA extracellular domain (aa 2-54).
- Tested applications:Functional Assay
- Purification:purified by Protein G affinity chromatography
- Pk:100 µG
BCMA (B cell maturation) is a nonglycosylated integral membrane type I protein that is preferentially expressed in mature B lymphocytes and belongs structurally to the TNF receptor family. BCMA associates with BAFF, APRIL, TNF receptor-associated factor (TRAF) 1, TRAF2, and TRAF3 and activates NF-κB, Elk-1, c-Jun N-terminal kinase (JNK), and p38 mitogen-activated protein kinase. BCMA plays an important role in B cell development, function and regulation.
Type: Primary
Antigen: BCMA
Clonality: Monoclonal
Clone: Vicky-1
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human