Specifications
- Antibody type:Primary
- Antigen name:Heat Shock Protein 60
- Antigen symbol:HSP60
- Clonality:Monoclonal
- Clone:LK-2
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Dog,Rat,Mouse,Hamster,Plant,Pig,Rabbit,Monkey,Bacteria,Chicken,C. elegans,Human,Bovine,Guinea pig
- Western blot:Yes
- Seq:02
- Form:Liquid
- Antigen synonyms:Mytonin Chaperone 60|GROEL|HSP65|P60 lymphocyte protein|HLD4|HSP-60|HuCHA60|mitochondrial matrix protein P1|chaperonin 60|HSP60|mitochondrial|CPN60|short heat shock protein 60 Hsp60s1|heat shock protein 65|60 kDa chaperonin|60 kDa heat shock protein|SPG13
- Storage buffer:PBS, pH 7,2, containing 0,1 mM PMSF and 50% glycerol
- Storage temperature:–20 °C
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant human HSP60
- Purification:Purified by Protein G affinity chromatography
- Pk:100 µG
Specifications
About this item
Hsp60 is a member of the chaperonin family of heat shock proteins, with homologs functioning in the cytosol and mitochondria to fold nascent and aggregated proteins. Hsp60 is the eukaryotic homolog of the E. coli GroEL protein, and forms a multimeric complex in the mitochondria with Hsp10 (Cpn10) to form a large central cavity in which ATP-dependent protein folding takes place. TRiC/CCT, a eukaryotic relative of Hsp60, is expressed in the cytosol and participates in the folding of actin and tubulin substrates, but lacks any association with an Hsp10-like co-factor.
Type: Primary
Antigen: HSP60
Clonality: Monoclonal
Clone: LK-2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Rabbit, Plant, Pig, Monkey, Hamster, Guinea pig, Dog, Chicken, C. elegans, Bovine, Bacteria