Anti-HSP47 Mouse Monoclonal Antibody [clone: M16.10A1]
Catalog # ENZOADISPA470J
Supplier: ENZO LIFE SCIENCES
Specifications
- Antibody type:Primary
- Antigen symbol:HSP47
- Clonality:Monoclonal
- Clone:M16.10A1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Hamster,Mouse,Pig,Rabbit,Monkey,Sheep,Chicken,Human,Bovine,Guinea pig,Dog,Rat
- Western blot:Yes
- Seq:02
- Form:Liquid
- Storage temperature:−20 °C
- Shipping temperature:Shipped on blue ice
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: HSP47
Clonality: Monoclonal
Clone: M16.10A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat, Sheep, Rabbit, Pig, Monkey, Hamster, Guinea pig, Dog, Chicken, Bovine