Anti-S Rabbit Polyclonal Antibody
ENZOADIKAPST210E
- Antibody type:Primary
- Antigen name:Serine
- Antigen symbol:Ser
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Multiple
- Western blot:Yes
- Seq:02
- Form:Liquid
- Antigen synonyms:Spike glycoprotein|Phosphoserine|Ser|E2|pSER|pS|SARS Spike|S glycoprotein|p-S|p-SER.
- Storage buffer:PBS containing 50% glycerol and 0,01% sodium azide
- Storage temperature:−20 °C
- Shipping temperature:Shipped on blue ice
- Immunogen:Phosphoserine conjugated to KLH
- Purification:Purified by peptide affinity chromatography
- Pk:100 µG
Protein phosphorylation is a complex biochemical process that is regulated by protein kinases that catalyze the transfer of the gamma-P of ATP to a recipient protein that acts as a substrate. This post-translational modification is primarily directed onto serine, threonine, and tyrosine amino acid residues within a protein. Changes in the phosphorylation status of phosphoproteins can alter their biological function, and aberrations in signal transduction cascades have been linked to a number of diseases, including cancer, diabetes, heart disease, inflammation and neurological disorders.
Type: Primary
Antigen: S
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: pan