Anti-BAFFR Rabbit Polyclonal Antibody
Catalog # ENZOADI905305100
Supplier: ENZO LIFE SCIENCES
Specifications
- Antibody type:Primary
- Antigen name:B-cell activating factor receptor
- Antigen symbol:BAFFR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Seq:02
- Epitope:near the C-Terminus
- Form:Liquid
- Storage buffer:PBS containing 0,02% sodium azide.
- Storage temperature:4 °C
- Shipping temperature:Blue ice not frozen
- Immunogen:Synthetic peptide corresponding to the sequence near the C-terminus of human BAFF receptor.
- Purification:purified by peptide affinity chromatography
- Pk:100 µG
Specifications
About this item
BAFF, or B-cell Activating Factor, is a member of the TNF superfamily also known as BLyS (B Lymphocyte Stimulator), TALL-1 (TNF- and ApoLrelated Leukocyte-expressed Ligand), and THANK (TNF Homologue that Activate Apoptosis, NF-κB and c-jun N-terminal Kinase). BAFF/BLyS was characterized as a B-cell activator since it induced B-cell proliferation and immunoglobulin secretion. Three receptors for BAFF were identified and designated TACI, BCMA, and BAFF-R. BAFF and its receptors are essential for B-cell development, survival, and humoral immune responses.
Type: Primary
Antigen: BAFFR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-Terminus
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat