Order Entry
Slovenia
ContactUsLinkComponent
Anti-APMAP Mouse Monoclonal Antibody
Anti-APMAP Mouse Monoclonal Antibody
Catalog # ENZOALX804144C100
Supplier:  ENZO LIFE SCIENCES
Anti-APMAP Mouse Monoclonal Antibody
Catalog # ENZOALX804144C100
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Adipocyte Plasma Membrane Associated Protein
  • Antigen symbol:
    APMAP
  • Clonality:
    Monoclonal
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid. In PBS containing 0,02% sodium azide.
  • Antigen synonyms:
    a|Adipose most abundant gene transcript 1|30 kDa adipocyte complement-related protein|Adipocyte|ACRP30|C1q and collagen domain-containing protein
  • Concentration:
    1 mg/ml
  • Purification:
    ≥95% (SDS-PAGE).
  • Pk:
    100 µG

Specifications

About this item

Host: Mouse, Isotype: IgG1

  • For ELISA, Flow Cytometry, Immunoprecipitation, Western Blot

Recognizes human ACRP30headless. Does not cross-react with mouse ACRP30headless. Does not cross-react with full length adiponectin.

Type: Primary
Antigen: APMAP
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human