Order Entry
Slovenia
ContactUsLinkComponent
Anti-RIPK2 Rat Monoclonal Antibody
Anti-RIPK2 Rat Monoclonal Antibody
Catalog # ENZOALX804139C100
Supplier:  ENZO LIFE SCIENCES
Anti-RIPK2 Rat Monoclonal Antibody
Catalog # ENZOALX804139C100
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Receptor-interacting serine/threonine-protein kinase 2
  • Antigen symbol:
    RIPK2
  • Clonality:
    Monoclonal
  • Host:
    Rat
  • Isotype:
    IgG2a
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid. In PBS containing 0,02% sodium azide.
  • Antigen synonyms:
    CARD3|RICK|CARDIAK|receptor interacting serine/threonine kinase 2|GIG30|CCK|RIP2
  • Concentration:
    1 mg/ml
  • Pk:
    100 µG

Specifications

About this item

Host: Rat, Isotype: IgG2a

  • For Western Blot

Recognizes human RIP2.

Type: Primary
Antigen: RIPK2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human