Anti-PRF1 Mouse Monoclonal Antibody
ENZOALX804135C100
- Antibody type:Primary
- Antigen name:Perforin
- Antigen symbol:PRF1
- Clonality:Monoclonal
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Western blot:Yes
- Seq:02
- Form:Liquid. In PBS containing 0,02% sodium azide.
- Antigen synonyms:lymphocyte pore-forming protein|cytolysin|lymphocyte pore forming protein|perforin-1
- Concentration:1 mg/ml
- Pk:100 µG
Host: Mouse, Isotype: IgG2a
- For Immunoprecipitation, Western Blot
Recognizes an epitope in the region aa 402-534 of human and mouse perforin.
Type: Primary
Antigen: PRF1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human