Anti-GRP75 Mouse Monoclonal Antibody
Catalog # ENZOALX804077R100
Supplier: ENZO LIFE SCIENCES
Specifications
- Antibody type:Primary
- Antigen name:Glucose-regulated Protein 75kDa
- Antigen symbol:GRP75
- Clonality:Monoclonal
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG3
- Reactivity:Human
- Western blot:Yes
- Seq:02
- Form:Liquid. Ascites diluted in PBS containing 0,05% sodium azide.
- Pk:100 µl
Specifications
About this item
Host: Mouse, Isotype: IgG3
- For Immunocytochemistry, Immunoprecipitation, Western Blot
Recognizes human, mouse, dog and monkey mtHSP70. Detects a band of ~75kDa by Western blot.
Type: Primary
Antigen: GRP75
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human