Anti-CASP3 Rabbit Polyclonal Antibody
Catalog # ENZOALX210807C100
Supplier: ENZO LIFE SCIENCES
Specifications
- Antibody type:Primary
- Antigen name:Caspase 3
- Antigen symbol:CASP3
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Seq:02
- Form:Liquid. In PBS containing 30% glycerol, 0,5% BSA and 0,01% thimerosal.
- Antigen synonyms:caspase-3|procaspase3|caspase 3|protein Yama|CPP-32,PARP cleavage protease|Yama|cysteine protease CPP32|CASP-3|apoptosis-related cysteine protease|apopain|SREBP cleavage activity 1
- Concentration:0,2 mg/ml
- Purification:Epitope-affinity purified
- Pk:100 µG
Specifications
About this item
Host: Rabbit
- For Immunohistochemistry, Western Blot
Recognizes human, mouse and rat active caspase-3 (p20 subunit). Does not cross-react with procaspase-3 (~32kDa).
Type: Primary
Antigen: CASP3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human