Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-MMP2 Rabbit Polyclonal Antibody
  ENZOALX210753R500
Anti-MMP2 Rabbit Polyclonal Antibody
  ENZOALX210753R500

 

  • Antibody type:
    Primary
  • Antigen name:
    Matrix Metalloproteinase 2
  • Antigen symbol:
    MMP2
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid. Neat serum.
  • Antigen synonyms:
    MMP-2|72 kDa gelatinase|matrix metalloproteinase-2|collagenase type IV-A|neutrophil gelatinase|gelatinase A|matrix metalloproteinase-II|72 kDa type IV collagenase
  • Pk:
    500 µl

 

 

Host: Rabbit

  • For Immunoprecipitation, Western Blot

Recognizes human and marmoset MMP-2. Moderately cross-reacts with MMP-1. Does not cross-react with MMP-3, MMP-8 or MMP-9.

Type: Primary
Antigen: MMP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human