Order Entry
Slovenia
ContactUsLinkComponent
Anti-Glucocorticoid Receptor Alpha Rabbit Polyclonal Antibody
Anti-Glucocorticoid Receptor Alpha Rabbit Polyclonal Antibody
Catalog # ENZOALX210277C050
Supplier:  ENZO LIFE SCIENCES
Anti-Glucocorticoid Receptor Alpha Rabbit Polyclonal Antibody
Catalog # ENZOALX210277C050
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Glucocorticoid Receptor Alpha
  • Antigen symbol:
    GCRA
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Seq:
    02
  • ChIP:
    Yes
  • Form:
    Lyophilised. Contains 0,05% sodium azide.
  • Purification:
    Epitope-affinity purified IgG
  • Pk:
    50 µG

Specifications

About this item

Host: Rabbit

  • For Immunocytochemistry, Immunoprecipitation, Western Blot, ChIP

Recognizes human, mouse and rat GRalpha. Detects a band of ~94kDa by Western blot.

Reconstitute with PBS.

Type: Primary
Antigen: GLUCOCORTICOID RECEPTOR ALPHA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human