ContactUsLinkComponent
Anti-DNAK Mouse Monoclonal Antibody [clone: 8E2/2]
undefined
Anti-DNAK Mouse Monoclonal Antibody [clone: 8E2/2]
  • Catalog No:
  • ENZOADISPA880J
  • ENZOADISPA880F
  • ENZOADISPA880D
  • Antigen symbol:
  • DNAK
  • DNAK
  • DNAK
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Clone:
  • 8E2/2
  • 8E2/2
  • 8E2/2
  • Reactivity:
  • Bacteria
  • Bacteria
  • Bacteria
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Liquid
  • Purification:
  • Protein G affinity purified
  • Protein G affinity purified
  • Protein G affinity purified
  • Storage buffer:
  • PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide
  • PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide
  • PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide
Frequently Bought Together