Anti-MAPKK5 Rabbit Polyclonal Antibody
ENZOADIKAPMA003E
- Antibody type:Primary
- Antigen name:Mitogen Activated Protein Kinase Kinase 5
- Antigen symbol:MAPKK5
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Seq:02
- Antigen synonyms:MAP kinase kinase 5|dual specificity mitogen-activated protein kinase kinase 5|PRKMK5|MEK5|MEK 5|MAPKK 5|MAP kinase kinase MEK5b|MAPKK5|HsT17454|MAPK/ERK kinase 5
- Size:100 µg
- Pk:100 µG
MEK is a dual specificity kinase capable of phosphorylating both tyrosine and threonine residues. The MEK family, which is also known as MAP kinase kinase, phosphorylates MAP kinases on the conserved T-X-Y motif; phosphorylation of MAPK by MEK results in an increase in MAPK activity. MEK is involved in a diverse array of cellular processes such as stress-activated response, apoptosis, cytokine-induced cell proliferation, and DNA recombination during meiosis.
- For Immunohistochemistry, Western Blot
Recognizes human, mouse, rat, dog, monkey, rabbit, sheep and Xenopus MEK5. Detects a band of ~42kDa by Western blot.
Type: Primary
Antigen: MAPKK5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human