Anti-ATLF Mouse Monoclonal Antibody
ENZOADI905621100
- Antibody type:Primary
- Antigen name:Anthrax Toxin Lethal Factor
- Antigen symbol:ATLF
- Clonality:Monoclonal
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Bacteria
- Western blot:Yes
- Seq:02
- Form:Liquid. In PBS containing 0,01% sodium azide. Concentration 1 mg/ml.
- Antigen synonyms:Anthrax lethal factor|Anthrax lethal toxin endopeptidase component|LF|Anthrax LF|LEF_BACAN.|bacillus anthracis lethal factor|Lef
- Purification:Protein G affinity purified
- Pk:100 µG
Anthrax Lethal Factor (LF) is a zinc-dependent protease that cleaves mitogen-activated protein kinase kinase (MAPKK) and causes lysis of macrophages. LF, together with Protective Antigen, forms Lethal Toxin, which is the dominant virulence factor produced by B. anthracis and is the major cause of death in infected animals. Lethal toxin has been reported to trigger apoptosis in human melanoma cells.
- For ELISA, Flow Cytometry, Immunoprecipitation, Western Blot
Recognizes bacteria Anthrax Lethal Factor (LF).
Type: Primary
Antigen: ATLF
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bacteria