Orders LinkContactUsLinkComponent
Sight Boxes, PP
Sight Boxes, PP
Supplier:   SEMADENI
Sight Boxes, PP
Supplier:  SEMADENI
Certificates
3 Options Available Below
Product Details & Documents
Made of PP.
  • Cost-effective
  • Resistant to greases, resins, alkalis, oils and acids
  • Good temperature resistance
These storage boxes are for the professional sector and a wide range of applications in industry, crafts, household and hobby.
Product Family Options
Product Information
  • Colour
    W×D×H
    Availability
    Price
  • Red
    210×240×145 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 7,50 €
    7,50 €
    1 * 1 items
       7,50 €
  • Blue
    210×240×145 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 7,50 €
    7,50 €
    1 * 1 items
       7,50 €
  • Red
    495/460×31×185 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 33,80 €
    33,80 €
    1 * 1 items
       33,80 €