ContactUsLinkComponent
Bottles, PE, rectangular, with screw cap
Bottles, PE, rectangular, with screw cap
Catalog # LAMA635
Supplier:  LAMAPLAST
Bottles, PE, rectangular, with screw cap
Catalog # LAMA635
Supplier:  LAMAPLAST
Specifications
  • Capacity:
    1000 ml
  • Colour:
    White
  • W×D×H:
    70×90×225 mm
  • Cap colour:
    White
  • Neck-Ø:
    35 mm
  • Pk:
    1
Specifications
About this item
This HDPE graduated bottle is suitable for the storage of dense fluids, granulates and powders.
  • Screw cap with opening-off seal
  • For food use
  • Emptying guaranted by the inclination of the shoulders