Orders LinkContactUsLinkComponent
Bottle brushes, with head bundle
Bottle brushes, with head bundle
Certificates
Bottle brushes, with head bundle
7 Options Available Below
Product Details & Documents
The brush consists of galvanised rotary wires with head bundle.
Product Family Options
Product Information
  • Bristle material
    Brush head length
    Brush head Ø
    Handle material
    Total length
    Availability
    Price
  • REIT21500234
    Brush for 0,25 l bottle, with bleached bristle fill material
    Brush for 0,25 l bottle, with bleached bristle fill material
    Hog bristle
    85 mm
    50 mm
    Steel, zinc plated
    390 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 88,90 €
    88,90 €
    1 * 10 items
       88,90 €
  • REIT22550234
    Brush for 0,50 l bottle, with bleached bristle fill material
    Brush for 0,50 l bottle, with bleached bristle fill material
    Hog bristle
    100 mm
    55 mm
    Steel, zinc plated
    420 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 101,00 €
    101,00 €
    1 * 10 items
       101,00 €
  • REIT23630202
    Brush for 0,75 l bottle, with horsehair black fill material
    Brush for 0,75 l bottle, with horsehair black fill material
    Horse hair
    120 mm
    63 mm
    Steel, zinc plated
    470 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 126,00 €
    126,00 €
    1 * 10 items
       126,00 €
  • REIT23630203
    Brush for 0,75 l bottle, with nylon fill material
    Brush for 0,75 l bottle, with nylon fill material
    Nylon
    120 mm
    63 mm
    Steel, zinc plated
    470 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 103,00 €
    103,00 €
    1 * 10 items
       103,00 €
  • REIT63800202
    Brush for 1 l bottle, with Horsehair black fill material
    Brush for 1 l bottle, with Horsehair black fill material
    Horse hair
    140 mm
    80 mm
    Steel, zinc plated
    550 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 167,00 €
    167,00 €
    1 * 10 items
       167,00 €
  • REIT63800203
    Brush for 1 l bottle, with nylon fill material
    Brush for 1 l bottle, with nylon fill material
    Nylon
    140 mm
    80 mm
    Steel, zinc plated
    550 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 113,00 €
    113,00 €
    1 * 10 items
       113,00 €
  • REIT63800234
    Brush for 1 l bottle, with bleached bristle fill material
    Brush for 1 l bottle, with bleached bristle fill material
    Hog bristle
    140 mm
    80 mm
    Steel, zinc plated
    550 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 174,00 €
    174,00 €
    1 * 10 items
       174,00 €