ContactUsLinkComponent
Disposable particle filtering respirators, P100
Disposable particle filtering respirators, P100
Catalog # PRTWP100WHR
Supplier:  Portwest
undefined
Disposable particle filtering respirators, P100
Catalog # PRTWP100WHR
Supplier:  Portwest
Specifications
  • Type:
    Particle filter
  • Pk:
    20
  • With valve:
    -
  • Class:
    FFP1
Specifications
About this item
This disposable respirator is designed for applications involving dust, mists and fibres. The P100 can be used in concentrations of contaminants up to 4 times the WEL.
  • Soft foam nose seal
  • Elasticated twin straps
  • Adjustable nose bridge