Orders LinkContactUsLinkComponent
Connection stopper
Connection stopper
Connection stopper for distribution heads with connection pipe Ø 32,7 mm
Catalog # 458-2357
Supplier:   GERHARDT
Connection stopper
Catalog # 458-2357
Supplier:  GERHARDT
Connection stopper
Connection stopper for distribution heads with connection pipe Ø 32,7 mm
Price
63,25 €
Log in for availability
1 * 1 items
Product Details & Documents
This connection stopper is made from viton and is suitable for distribution heads with connection pipe Ø 32,7 mm.
Specifications
  • Description:
    Connection stopper for distribution heads with connection pipe Ø 32,7 mm