Standards for water pollution QC check, inorganics, VHG
Catalog # PROOVHG-QWSIN-500
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:500 mL
- Description:Water analysis standard for inorganics, 500 ml
- Type:Reference standard
Specifications
About this item
This solution is intended for use as a quality control check for methods related to measurements of inorganic materials includes conductivity, alkalinity as CaCO₃, chloride, fluoride, nitrite and nitrate, sulphate, potassium, sodium, total dissolved solids and many more.
This standard has been prepared as a whole volume standard and is ready to be analysed as received. This standard is not preserved.
Certifications: Traceable to NIST SRM.