Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
Forceps
Tissue forceps, Taylor
Tissue forceps, Taylor
232-2177
:
HAMMACHER KARL
Tissue forceps, Taylor
232-2177
:
HAMMACHER KARL
Type:
Straight
Description:
Taylor, delicate
Length:
180 mm
Seq:
07
Pk:
1
Delicate forceps made from stainless steel.