Orders LinkContactUsLinkComponent
Mailing boxes, mini
Mailing boxes, mini
Mailing box, white, 240×129×30 mm, 120×230×28 mm
Catalog # DAKL20121
Supplier:   DAKLAPACK
Mailing boxes, mini
Catalog # DAKL20121
Supplier:  DAKLAPACK
Mailing boxes, mini
Mailing boxes, mini
Mailing box, white, 240×129×30 mm, 120×230×28 mm
Price
134,00 €
Log in for availability
1 * 100 items
Product Details & Documents
Mini mailing boxes are designed for sending diagnostic samples of Category B.
  • Cardboard material printed with UN-number and logo as per regulations
Certifications: Approved (BAM and postal organisations) as external packaging for diagnostisc samples of Category B, as long as used in accordance with packaging instruction P650 in combination with safetybag and absorbent tube holders.
Documents
Specifications
  • Description:
    Mailing box, white
  • W×D×H ext.:
    240×129×30 mm
  • W×D×H int.:
    120×230×28 mm