Orders LinkContactUsLinkComponent
Autoclavable goggles
 :   UNIVET
Autoclavable goggles
 :  UNIVET
Autoclavable goggles
Autoclavable goggles
The autoclavable goggles are designed for use in rooms with controlled contaminations or in aseptic conditions.
  • Available with direct or indirect ventilation
  • Elastic band in anti-static silicone with quick and smooth system
  • RX insert
  • Goggles with polycarbonate lens reusable up to 40 autoclave
The soft structure supports multiple cycles of steam, gamma or beta sterilisation without perceptible changes. The autoclavable goggles has a polycarbonate lens with anti-fog coating AF121 and anti-scratch.
 : EN 166.
 : Available with correction insert and replacement lens.
Product Family Options
  • Description
    Type
    Lenses
    Colour
  • Cleanroom goggles
    Directly vented
    Clear βγoxene® disc
    Blue
    1 * 5 items
    1 * 5 items - 293,00 €
    293,00 €
    1 * 5 items
       293,00 €
  • Cleanroom goggles
    Anti-UV, anti-fog, autoclavable, directly vented
    Clear PC
    Blue
    1 * 5 items
    1 * 5 items - 357,00 €
    357,00 €
    1 * 5 items
       357,00 €
  • Cleanroom goggles
    Anti-UV, autoclavable, indirectly vented
    Clear PC
    Blue
    1 * 5 items
    1 * 5 items - 293,00 €
    293,00 €
    1 * 5 items
       293,00 €
  • Cleanroom goggles
    Anti-UV, anti-fog, autoclavable, indirectly vented
    Clear PC
    Blue
    1 * 5 items
    1 * 5 items - 366,00 €
    366,00 €
    1 * 5 items
       366,00 €
Accessories
  •  
    Description
  • UNVT042.060
    Spare lens for 611S, clear PC
    1 * 1 items - 14,80 €
  •  
    Description
  • UNVT3PA299
    Head band, silicone, 700×30×1,4 mm
    1 * 1 items - 19,60 €
  •  
    Description
  • UNVT601.OP.00.99
    RX optical insert for goggles
    1 * 1 items - 14,30 €
  •  
    Description
  • UNVT042.120M
    Spare lens, clear PC
    1 * 1 items - 33,40 €