ContactUsLinkComponent
Portable padlock station
Portable padlock station
Catalog # BRDY811218
Supplier:  Brady
undefined
Portable padlock station
Catalog # BRDY811218
Supplier:  Brady
Specifications
  • Type:
    Portable padlock station
  • Colour:
    Yellow
  • W×D×H:
    337×64×432 mm
  • Pk:
    1
Specifications
About this item
This portable storage station ensures that lockout devices remain easily accessible and ready for use whenever needed.
  • Made from rugged and rust-proof PP
  • Attachable handle for easy portability
  • Screw slots for permanent wall mounting
  • Lockable, semi-transparent cover protects the contents from exposure to harsh environments
Ordering information: Station is supplied empty (without contents).