Elemental analysis standards, phosphate rock, western, NIST-694
Catalog # PROONIST-694
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:90 g
- Description:NIST-694, western phosphate rock
Specifications
About this item
This standard material is intended to use in validating analytical methods and in calibrating instruments used in the analysis of phosphate rock, fertilisers and similar materials.
A unit consists of 90 g of <150 µm (100 mesh) powdered material.
Certifications: Traceable to NIST and SRM.