ContactUsLinkComponent
Mouse recombinant CD279 (from CHO cells)
Mouse recombinant CD279 (from CHO cells)
Catalog # PRSI90-449
Supplier:  ProSci Inc.
CAS Number:  
Mouse recombinant CD279 (from CHO cells)
Catalog # PRSI90-449
Supplier:  ProSci Inc.
CAS Number:  
Specifications
  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Mouse
  • Tag sequence:
    Fc Tag
  • Storage conditions:
    Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
  • Protein synonyms:
    PD-1|Programmed Cell Death Protein 1
  • Protein/peptide name:
    CD279
  • Purity:
    >98% (SDS-PAGE)
  • Sequence:
    26-169
  • Formulation:
    Lyophilized from 0.2um-filtered solution in PBS.
Specifications
About this item
CD279 (Programmed Cell Death Protein 1; PD-1) is a type I transmembrane protein belonging to the CD28/CTLA-4 family of immunoreceptors that mediate signals for regulating immune responses. Members of the CD28/CTLA-4 family have been shown to either promote T cell activation (CD28 and ICOS) or downregulate T cell activation (CTLA-4 and PD-1). CD279 is expressed on activated T cells, B cells, myeloid cells and on a subset of thymocytes. In vitro, ligation of CD279 inhibits TCR-mediated T cell proliferation and production of IL-1, IL-4, IL-10 and IFN-gamma. In addition, CD279 ligation also inhibits BCR mediated signallling. CD279 deficient mice have a defect in peripheral tolerance and spontaneously develop autoimmune diseases.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.