Order Entry
Slovenia
ContactUsLinkComponent
Mouse recombinant CD273 (from CHO cells)
Mouse recombinant CD273 (from CHO cells)
Catalog # PRSI90-428
Supplier:  ProSci Inc.
CAS Number:  
Mouse recombinant CD273 (from CHO cells)
Catalog # PRSI90-428
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Mouse
  • Tag sequence:
    Fc Tag
  • Storage conditions:
    Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
  • Protein synonyms:
    B7-CD|Programmed Cell Death 1 Ligand 2|PDL2|PDCD1 Ligand 2
  • Protein/peptide name:
    CD273
  • Purity:
    >98% (SDS-PAGE)
  • Sequence:
    20-219
  • Formulation:
    Lyophilized from 0.2um-filtered solution in PBS.

Specifications

About this item

T cells require a signal induced by the engagement of the T cell receptor and a costimulatory signal(s) through distinct T cell surface molecules for optimal T cell activation and tolerance. CD273 (PD-L2) is one of two ligands for programmed death-1 (PD-1; CD279), a member of the CD28 family of immunoreceptors. The other identified ligand is PD-L1. CD273 is broadly expressed and also up regulated in a variety of tumour cell lines. On previously activated T cells, CD273 interaction with PD-1 inhibits TCR mediated proliferation and cytokine production, suggesting an inhibitory role in regulating immune responses. CD273 has a costimulatory function on resting T cells activated with suboptimal TCR signals.

  • This recombinant proteins is for research use only.

Fusion-Tag: Fc Tag

This recombinant proteins is for research use only.