Order Entry
Slovenia
ContactUsLinkComponent
Cynomolgus monkey recombinant PDL12 (from HEK293 cells)
Cynomolgus monkey recombinant PDL12 (from HEK293 cells)
Catalog # PRSI96-592
Supplier:  ProSci Inc.
CAS Number:  
undefined
Cynomolgus monkey recombinant PDL12 (from HEK293 cells)
Catalog # PRSI96-592
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Cynomolgus monkey
  • Protein synonyms:
    B7-H1|PD-L1|CD274|PDCD1L1|PDCD1LG1
  • Protein/peptide name:
    PDL12
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    51.8 kDa
  • Formulation:
    Lyophilized, 50 mM tris, 100 mM glycine, pH7.5
  • Tested applications:
    Western Blot

Specifications

About this item

Programmed cell death 1 ligand 1 (PDL1) is also known as B7-H, B7H1, MGC142294, MGC142296, PD-L1, PDCD1L1 and PDCD1LG1,which is a member of the growing B7 family of immune molecules and is involved in the regulation of cellular and humoral immune responses.PDL1 is a cell surface immunoglobulin superfamily with two Ig-like domains within the extracellular region and a short cytoplasmic domain. This protein is broadly expressed in the majority of peripheral tissues as well as hematopoietic cells. Interaction between PDL1 and its receptors belonging to the CD28 family of molecules provide both stimulatory and inhibitory signals in regulating T cell activation and tolerance. PDL1 may inhibit ongoing T-cell responses by inducing apoptosis and arresting cell-cycle progression.