Order Entry
Slovenia
ContactUsLinkComponent
Human Recombinant CD14 (from HEK293 cells)
Human Recombinant CD14 (from HEK293 cells)
Catalog # PRSI96-111
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human Recombinant CD14 (from HEK293 cells)
Catalog # PRSI96-111
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Protein/peptide name:
    CD14
  • Purity:
    >97% as determined by SDS-PAGE.
  • Molecular weight:
    62.4 kDa
  • Formulation:
    Lyophilized, 50 mM tris, 100 mM glycine, pH7.5
  • Tested applications:
    Western Blot

Specifications

About this item

Cluster of differentiation 14 (CD14), is a cell surface glycoprotein, and is a is a component of the innate immune system. CD14 is a myelomonocytic differentiation antigen preferentially expressed on monocytes, macrophages, and activated granulocytes. CD14 exists in two forms. Either it is anchored into the membrane by a glycosylphosphatidylinositol tail (mCD14) or it appears in a soluble form (sCD14). Soluble CD14 either appears after shedding of mCD14 (48 kDa) or is directly secreted from intracellular vesicles (56 kDa). CD14 acts as a co-receptor (along with the Toll-like receptor TLR 4 and MD-2) for the detection of bacterial lipopolysaccharide (LPS). CD14 can bind LPS only in the presence of lipopolysaccharide-binding protein (LBP). CD14 has been proposed to be involved in various biological processes, including transportation of other lipids, cell-cell interaction during different immune responses, as well as recognition of apoptotic cells. Although LPS is considered its main ligand, CD14 also recognizes other pathogen-associated molecular patterns. CD14+ cells are monocytes that can differentiate into a host of different cells. CD14 has been shown to interact with Lipopolysaccharide-binding protein.