Anti-POLG Rabbit Polyclonal Antibody
Catalog # PRSI71-303
Supplier: ProSci Inc.
Specifications
- Antigen name:Polymerase (DNA) Gamma, Catalytic Subunit
- Antigen symbol:POLG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:MTDPS4B|MTDPS4A|SANDO|POLG1|MDP1|PEO|MIRAS|SCAE|POLGA
- Concentration:0.7 mg/mL
- Immunogen:Recombinant protein fragment containing a sequence corresponding to a region within amino acids 829 and 1201 of DNA polymerase gamma.
- Tested applications:Immunoaffinity Chromatography
- Size:50 µl
- Pk:50 µl
Specifications
About this item
POLG Antibody can be used for Immunocytochemistry starting at 1:100 - 1:200, Immunofluorescence starting at 1:100 - 1:200, Immunohistochemistry in paraffin embeded tissues starting at 10μg/mL, and Western Blotting starting at 1:1000 - 1:10000.
PBS, pH 7, 1% BSA, 20% Glycerol, 0.01% Thimerosal
PBS, pH 7, 1% BSA, 20% Glycerol, 0.01% Thimerosal
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: POLG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human