Order Entry
Slovenia
ContactUsLinkComponent
Anti-BRSK1 Rabbit Polyclonal Antibody
Anti-BRSK1 Rabbit Polyclonal Antibody
Catalog # PRSI4081
Supplier:  ProSci Inc.
undefined
Anti-BRSK1 Rabbit Polyclonal Antibody
Catalog # PRSI4081
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    BR serine/threonine kinase 1
  • Antigen symbol:
    BRSK1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Antigen synonyms:
    SAD1|KIAA1811|Brain-selective kinase 1|SADB|SAD1 homolog|hSAD1
  • Storage buffer:
    PBS containing 0.02% sodium azide
  • Immunogen:
    BRSK1 antibody was raised against a 28 amino acid synthetic peptide from near the carboxy terminus of human BRSK1.
  • Tested applications:
    E, WB, IHC, IF
  • Purification:
    Affinity chromatography purified via peptide column
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

BRSK1 Antibody: BRSK1 was initially identified as a mammalian homolog to the fission yeast S. pombe Cdr2, a mitosis-regulatory kinase and also shows significant homology to the C. elegans neuronal cell polarity regulator SAD1. BRSK1 is unbiquitously expressed, with highest levels of expression in the brain and testes. Similar to its yeast homolog, BRSK1 is thought to be involved in stress-induced cell cycle arrest. Overexpression of this protein leads to the G2/M arrest in HeLa S2 cells and UV-induced G2/M arrest could be partially abrogated by reduced expression of BRSK1 through the use of siRNA, indicating its role in DNA damage checkpoint function. More recently, it has been shown that both BRSK1 and the related protein BRSK2 are required for mammalian neuronal polarization. While BRSK1- and BRSK2-null mice were viable, double-mutant mice died within two hours of birth. Neurons from these mice showed uniformly-sized neurites as opposed to the normal long axon and multiple shorter dendrites. These neurites also displayed both axonal and dendritic markers. At least two isoforms of BRSK1 are known to exist.

Type:
Antigen: BRSK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat