Orders LinkContactUsLinkComponent
Lockout tool boxes
Lockout tool boxes
Supplier:   Brady
Lockout tool boxes
Supplier:  Brady
Certificates
Lockout tool boxes
Lockout tool boxes
2 Options Available Below
Product Details & Documents
These lockout tool boxes are made of rugged polypropylene with nickel-plated steel clasps.
  • Two handy storage pockets built into lid
  • Removable tray for organising contents
Delivery information: Supplied with removeable tray and two handy storage pockets built into the lid.
Product Family Options
Product Information
  • Type
    Colour
    W×D×H
    Availability
    Price
  • Lockout tool box, small
    Black/red
    355,6×139,7×180,3 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 29,80 €
    29,80 €
    1 * 1 items
       29,80 €
  • Lockout tool box, medium
    Black/red
    411,5×185,4×198,1 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 37,20 €
    37,20 €
    1 * 1 items
       37,20 €