LoGiCal reference materials, (2-oxopyrrolidin-1-yl)acetic acid
PROOLGCFOR0083.01
:
- Pk:10 mg
- Standards:Reference standard
- CAS No.:53934-76-2
- Description:(2-Oxopyrrolidin-1-yl)acetic acid
- Formula:C₆H₉NO₃
- Description:(2-Oxopyrrolidin-1-yl)acetic acid
: Accredited to ISO 9001.