Capillary columns, Pora-PLOT Q
Supplier: Agilent
Certificates
About this item
These Pora-PLOT Q columns are mostly recommended for the column switching systems that involves in the analysis of polar and apolar volatile compounds.
- Available in fused silica and ultiMetal
- Long term stability provides repeatable retention times
- Water elutes as a sharp peak enabling quantitation
Each of these type of columns are tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency to provide you utmost confidence in qualitative and quantitative results.
Ordering information: Similar phases are available as Rt-Q BOND, Rt-QPLOT, SupelQ PLOT columns.
Frequently Bought Together